Anti-CD86 Rat Polyclonal Antibody [clone: GL1]

Lieferant: OriGene
TA320296
ORIGTA320296EA 314 EUR
ORIGTA320296
Anti-CD86 Rat Polyclonal Antibody [clone: GL1]
Antikörper
Type: Primary
Antigen: CD86
Clonality: Polyclonal
Clone: GL1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR