Anti-CD8A Mouse Polyclonal Antibody [clone: HIT8a]

Lieferant: OriGene
TA320300
ORIGTA320300EA 284 EUR
ORIGTA320300
Anti-CD8A Mouse Polyclonal Antibody [clone: HIT8a]
Antikörper
Type: Primary
Antigen: CD8A
Clonality: Polyclonal
Clone: HIT8a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR