Anti-IFNGR2 Mouse Polyclonal Antibody [clone: 4S.B3]

Lieferant: OriGene
TA320354
ORIGTA320354EA 245 EUR
ORIGTA320354
Anti-IFNGR2 Mouse Polyclonal Antibody [clone: 4S.B3]
Antikörper
Type: Primary
Antigen: IFNGR2
Clonality: Polyclonal
Clone: 4S.B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR