Anti-IL1B Armenian Hamster Polyclonal Antibody [clone: B122]

Lieferant: OriGene
TA320365
ORIGTA320365EA 284 EUR
ORIGTA320365
Anti-IL1B Armenian Hamster Polyclonal Antibody [clone: B122]
Antikörper
Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone: B122
Conjugation: Unconjugated
Epitope:
Host: Armenian Hamster
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR