Anti-LAT Mouse Polyclonal Antibody [clone: LAT.10-17 (10-17)]

Lieferant: OriGene
TA320411
ORIGTA320411EA 284 EUR
ORIGTA320411
Anti-LAT Mouse Polyclonal Antibody [clone: LAT.10-17 (10-17)]
Antikörper
Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone: LAT.10-17 (10-17)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR