Anti-OPRM1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA321234
ORIGTA321234EA 528 EUR
ORIGTA321234
Anti-OPRM1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: OPRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR