Anti-SLC15A2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA322236
ORIGTA322236EA 528 EUR
ORIGTA322236
Anti-SLC15A2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:2000-5000; Immunochemistry: 1:50-200

Type: Primary
Antigen: SLC15A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR