Anti-EPPIN Rabbit Polyclonal Antibody

Lieferant: OriGene
TA322256
ORIGTA322256EA 528 EUR
ORIGTA322256
Anti-EPPIN Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: EPPIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR