Anti-HIST1H3D Rabbit Polyclonal Antibody

Lieferant: OriGene
TA322846
ORIGTA322846EA 528 EUR
ORIGTA322846
Anti-HIST1H3D Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:100-200

Type: Primary
Antigen: HIST1H3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR