Anti-CASP1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA323204
ORIGTA323204EA 528 EUR
ORIGTA323204
Anti-CASP1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:1000-10000; Western Blot: 1:1000-4000; Immunochemistry: 1:25-100

Type: Primary
Antigen: CASP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR