Anti-BID Rabbit Polyclonal Antibody

Lieferant: OriGene
TA323207
ORIGTA323207EA 528 EUR
ORIGTA323207
Anti-BID Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:200-2000; Immunochemistry: 1:25-100

Type: Primary
Antigen: BID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR