Anti-TUBB3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA324074
ORIGTA324074EA 528 EUR
ORIGTA324074
Anti-TUBB3 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: ELISA: 1:500-5000; Western Blot: 1:500-1000; Immunochemistry: 1:25-50

Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR