Anti-CD22 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA324091
ORIGTA324091EA 528 EUR
ORIGTA324091
Anti-CD22 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: CD22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR