Anti-TAF9 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA324556
ORIGTA324556EA 805 EUR
ORIGTA324556
Anti-TAF9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:1000

Type: Primary
Antigen: TAF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR