Anti-TRIM25 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA324607
ORIGTA324607EA 805 EUR
ORIGTA324607
Anti-TRIM25 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TRIM25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR