Anti-ATF2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA324608
ORIGTA324608EA 805 EUR
ORIGTA324608
Anti-ATF2 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR