Anti-ZNF155 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA324612
ORIGTA324612EA 805 EUR
ORIGTA324612
Anti-ZNF155 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ZNF155
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR