Anti-STAT1 Mouse Monoclonal Antibody [clone: 1141CT26.2.1]

Lieferant: OriGene
TA324835
ORIGTA324835EA 805 EUR
ORIGTA324835
Anti-STAT1 Mouse Monoclonal Antibody [clone: 1141CT26.2.1]
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 1141CT26.2.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR