Anti-WNT10B Rabbit Polyclonal Antibody

Lieferant: OriGene
TA324853
ORIGTA324853EA 805 EUR
ORIGTA324853
Anti-WNT10B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: WNT10B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR