Anti-MAP3K6 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA325005
ORIGTA325005EA 805 EUR
ORIGTA325005
Anti-MAP3K6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: MAP3K6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR