Anti-MAP2K2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA325009
ORIGTA325009EA 805 EUR
ORIGTA325009
Anti-MAP2K2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR