Anti-AOC3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA325044
ORIGTA325044EA 805 EUR
ORIGTA325044
Anti-AOC3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: AOC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR