Anti-HDAC8 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA325525
ORIGTA325525EA 528 EUR
ORIGTA325525
Anti-HDAC8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: HDAC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR