Anti-ITGB3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA325580
ORIGTA325580EA 528 EUR
ORIGTA325580
Anti-ITGB3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: ITGB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR