Anti-SLC5A5 Mouse Monoclonal Antibody [clone: 14F]

Lieferant: OriGene
TA326382
ORIGTA326382EA 805 EUR
ORIGTA326382
Anti-SLC5A5 Mouse Monoclonal Antibody [clone: 14F]
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SLC5A5
Clonality: Monoclonal
Clone: 14F
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR