Anti-HspB8 Mouse Monoclonal Antibody [clone: 3C12-H11]

Lieferant: OriGene
TA326394
ORIGTA326394EA 792 EUR
ORIGTA326394
Anti-HspB8 Mouse Monoclonal Antibody [clone: 3C12-H11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HspB8
Clonality: Monoclonal
Clone: 3C12-H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR