Anti-LAT Mouse Monoclonal Antibody [clone: 11B.12]

Lieferant: OriGene
TA327925
ORIGTA327925EA 625 EUR
ORIGTA327925
Anti-LAT Mouse Monoclonal Antibody [clone: 11B.12]
Antikörper
Type: Primary
Antigen: LAT
Clonality: Monoclonal
Clone: 11B.12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR