Anti-SYK Mouse Monoclonal Antibody [clone: 4D10.2]

Lieferant: OriGene
TA327963
ORIGTA327963EA 750 EUR
ORIGTA327963
Anti-SYK Mouse Monoclonal Antibody [clone: 4D10.2]
Antikörper
Type: Primary
Antigen: SYK
Clonality: Monoclonal
Clone: 4D10.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR