Anti-CD247 Mouse Monoclonal Antibody [clone: 6B10.2]

Lieferant: OriGene
TA328059
ORIGTA328059EA 750 EUR
ORIGTA328059
Anti-CD247 Mouse Monoclonal Antibody [clone: 6B10.2]
Antikörper
Type: Primary
Antigen: CD247
Clonality: Monoclonal
Clone: 6B10.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 ?
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR