Anti-CSF1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA328372
ORIGTA328372EA 436 EUR
ORIGTA328372
Anti-CSF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CSF1
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR