Anti-GTF2F2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA329284
ORIGTA329284EA 674 EUR
ORIGTA329284
Anti-GTF2F2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR