Anti-SEZ6 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA337738
ORIGTA337738EA 749 EUR
ORIGTA337738
Anti-SEZ6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEZ6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR