Anti-AKT3 Mouse Monoclonal Antibody [clone: OTI9B2]

Lieferant: OriGene
TA500001
ORIGTA500001EA 621 EUR
ORIGTA500001
Anti-AKT3 Mouse Monoclonal Antibody [clone: OTI9B2]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AKT3 (v-akt murine thymoma viral oncogene homolog 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR