Anti-TRIM33 Mouse Monoclonal Antibody [clone: OTI9D4]

Lieferant: OriGene
TA500119
ORIGTA500119EA 621 EUR
ORIGTA500119
Anti-TRIM33 Mouse Monoclonal Antibody [clone: OTI9D4]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: TRIM33 (tripartite motif containing 33)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR