Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI4F1]

Lieferant: OriGene
TA500170
ORIGTA500170EA 621 EUR
ORIGTA500170
Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI4F1]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: CPA1 (carboxypeptidase A1 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR