Anti-BRAF Mouse Monoclonal Antibody [clone: OTI4B2]

Lieferant: OriGene
TA500438
ORIGTA500438EA 621 EUR
ORIGTA500438
Anti-BRAF Mouse Monoclonal Antibody [clone: OTI4B2]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR