Anti-BRAF Mouse monoclonal antibody [clone: OTI1G6]

Lieferant: OriGene
TA500439
ORIGTA500439EA 621 EUR
ORIGTA500439
Anti-BRAF Mouse monoclonal antibody [clone: OTI1G6]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR