Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI2C8]

Lieferant: OriGene
TA500457
ORIGTA500457EA 621 EUR
ORIGTA500457
Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI2C8]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1 (nuclear receptor binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR