Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI7C9]

Lieferant: OriGene
TA500473
ORIGTA500473EA 621 EUR
ORIGTA500473
Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI7C9]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Immunoprecipitation: 2-4μg/mg

Type: Primary
Antigen: MAPK1 (mitogen-activated protein kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR