Anti-KDM4C Mouse Monoclonal Antibody [clone: OTI4B1]

Lieferant: OriGene
TA500587
ORIGTA500587EA 621 EUR
ORIGTA500587
Anti-KDM4C Mouse Monoclonal Antibody [clone: OTI4B1]
Antikörper
Recommended Dilutions: Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KDM4C (lysine (K)-specific demethylase 4C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR