Anti-PPP5C Mouse Monoclonal Antibody [clone: OTI6C2]

Lieferant: OriGene
TA500596
ORIGTA500596EA 621 EUR
ORIGTA500596
Anti-PPP5C Mouse Monoclonal Antibody [clone: OTI6C2]
Antikörper
Recommended Dilutions: Western Blot: 1:250-500; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: PPP5C (protein phosphatase 5, catalytic subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR