Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI4G5]

Lieferant: OriGene
TA500598
ORIGTA500598EA 621 EUR
ORIGTA500598
Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI4G5]
Antikörper
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2μg/500μl

Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR