Anti-SLC7A8 Mouse Monoclonal Antibody [clone: OTI2D4]

Lieferant: OriGene
TA500631
ORIGTA500631EA 621 EUR
ORIGTA500631
Anti-SLC7A8 Mouse Monoclonal Antibody [clone: OTI2D4]
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SLC7A8 (solute carrier family 7 (amino acid transporter light chain, L system), member 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR