Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI3C11]

Lieferant: OriGene
TA500664
ORIGTA500664EA 621 EUR
ORIGTA500664
Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI3C11]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: LTA4H (leukotriene A4 hydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR