Anti-PSMA7 Mouse Monoclonal Antibody [clone: OTI8F9]

Lieferant: OriGene
TA500694
ORIGTA500694EA 621 EUR
ORIGTA500694
Anti-PSMA7 Mouse Monoclonal Antibody [clone: OTI8F9]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMA7 (proteasome (prosome, macropain) subunit, alpha type, 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR