Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI1G5]

Lieferant: OriGene
TA500711
ORIGTA500711EA 621 EUR
ORIGTA500711
Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI1G5]
Antikörper
Recommended Dilutions: Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B (ring finger protein 144B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR