Anti-PRL Mouse Monoclonal Antibody [clone: OTI2B7]

Lieferant: OriGene
TA500721
ORIGTA500721EA 621 EUR
ORIGTA500721
Anti-PRL Mouse Monoclonal Antibody [clone: OTI2B7]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PRL (prolactin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR