Anti-DPP4 Mouse Monoclonal Antibody [clone: OTI11D7]

Lieferant: OriGene
TA500733
ORIGTA500733EA 621 EUR
ORIGTA500733
Anti-DPP4 Mouse Monoclonal Antibody [clone: OTI11D7]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: DPP4 (dipeptidyl-peptidase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR