Anti-PPME1 Mouse Monoclonal Antibody [clone: OTI10B2]

Lieferant: OriGene
TA500769
ORIGTA500769EA 621 EUR
ORIGTA500769
Anti-PPME1 Mouse Monoclonal Antibody [clone: OTI10B2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PPME1 (protein phosphatase methylesterase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR