Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI3H6]

Lieferant: OriGene
TA500776
ORIGTA500776EA 621 EUR
ORIGTA500776
Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI3H6]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TTLL12 (tubulin tyrosine ligase-like family, member 12)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR